Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCAR/CD89 Rabbit anti-Human, Clone: 2, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | FCAR/CD89 |
---|---|
Clone | 2 |
Dilution | Sandwich ELISA Capture 1:250-1:2000 |
Applications | ELISA |
Classification | Monoclonal |
Description
FCAR/CD89 Monoclonal specifically detects FCAR/CD89 in Human samples. It is validated for Sandwich ELISA Capture.Specifications
FCAR/CD89 | |
Sandwich ELISA Capture 1:250-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
2204 | |
IgG | |
Protein A purified |
2 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor | |
Recombinant Human FCAR/CD89 Protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title