Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ FCAR/CD89 Recombinant Protein

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

$287.00 - $1025.00

Specifications

Formulation PBS (pH 7.4) containing 10% glycerol. containingno Preservative
Gene Alias CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Gene ID (Entrez) 2204
Gene Symbol FCAR
Name Recombinant FCAR/CD89 Protein
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB6021410UG View Documents Novus Biologicals™
NBP26021410UG
10μg Each for $287.00
Add to cart
 
NB6021450UG View Documents Novus Biologicals™
NBP26021450UG
50μg Each for $1,025.00
Add to cart
 
Description

Description

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 22-227 of Human FCAR/CD89 The Recombinant Human FCAR/CD89 Protein is derived from Baculovirus. The Recombinant Human FCAR/CD89 Protein has been validated for the following applications: SDS-Page.
Specifications

Specifications

PBS (pH 7.4) containing 10% glycerol. containingno Preservative
2204
Recombinant FCAR/CD89 Protein
Baculovirus
0.25mg/mL
SDS-Page
CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
FCAR
Recombinant Protein
RUO
< 1.0 EU per 1ug of protein (determined by LAL method)

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit