Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCAR Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  BiossSupplier Diversity Partner BS6586R

Catalog No. 50-198-7355


Add to cart

Description

Description

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene.
Specifications

Specifications

FCAR
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Fcar
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
2204
Fcar
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P24071
CD89; CTB-61M7.2; Fc alpha receptor; Fc fragment of IgA receptor; Fc fragment of IgA, receptor for; FcalphaRI; FCAR; IgA Fc receptor; immunoglobulin A Fc receptor; immunoglobulin A Fc receptor transcript variant deltaEC1 delta TM/C; Immunoglobulin alpha Fc receptor
KLH conjugated synthetic peptide derived from human FCAR.
Protein A
RUO
Antibody
Polyclonal
Human
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit