Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FceR1 alpha Chimeric Recombinant Rabbit Monoclonal Antibody (IB10), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548232

Encompass

Catalog No. PIMA548232


Add to Cart

Description

Description

Specificity: This antibody binds to human Fc epsilon receptor and competes with IgE binding.

FceR1 alpha Chimeric Recombinant Monoclonal antibody specifically detects FceR1 alpha Chimeric in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Inhibition Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FceR1 alpha Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
O54907
Rabbit
Affinity chromatography
RUO
21944
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Flow Cytometry, Immunocytochemistry, Inhibition Assays
IB10
Unconjugated
Tnfsf12
alpha polypeptide; fc epsilon r1; Fc epsilon receptor Ia; Fc epsilon RI alpha-chain; Fc fragment of IgE receptor Ia; Fc fragment of IgE, high affinity I, receptor for; alpha polypeptide; Fc IgE receptor, alpha polypeptide; Fc receptor, IgE, high affinity I, alpha polypeptide; Fce1a; Fc-epsilon RI-alpha; fcepsilonri; FCER1A; FcERI; Fcr-5; high affinity immunoglobulin epsilon receptor alpha-subunit; high affinity immunoglobulin epsilon receptor subunit alpha; IgE Fc receptor subunit alpha; Iger01; immunoglobulin E receptor, high-affinity, of mast cells, alpha polypeptide; RATIGER01; RP11-550P17.3
This antibody was raised by immunising SD rats with CD8-mTWEAK, a fusion of the human CD8alpha extracellular region to murine TWEAK.
200 μg
Primary
Mouse
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit