Learn More
FCER1 Mouse anti-Human, Clone: 9E1, Abnova™
Mouse monoclonal antibody raised against human FceR1.
Manufacturer:  Abnova Corporation MAB12566
Catalog No. 89-934-126
Add to cart
Description
Description
Specifications
Specifications
FCER1
9E1
Mouse monoclonal antibody raised against human FceR1.
In PBS, pH 7.4 (0.09% sodium azide).
Mouse
IgG2b
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA Flow Cytometry (1:25-1:50) Immunofluorescence Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
Human FceR1
Affinity purification
RUO
Primary
Human