Learn More
Abnova™ FCER1A 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00002205T02
Catalog No. 89-944-300
Add to cart
Specifications
Specifications
Western Blot
293T
NM_002001.2
pCMV-FCER1A full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
FCER1A 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 29.6kDa
Transient overexpression cell lysate was tested with anti-FCER1A antibody (Mfr. No. H00002205-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)