Learn More
FCER2 Mouse anti-Human, Clone: OTI9B6, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801830
Catalog No. 50-166-5741
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CD23 is a 45 kDa glycoprotein which is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. The CD23 molecule is identical to the low affinity IgE receptor found on B cells. Expression of CD23 has been detected in neoplastic cells from cases of B cell chronic lymphocyctic leukaemia and some cases of centroblastic/centrocytic lymphoma. CD23 is present on a subpopulation of freshly isolated peripheral blood and tonsil B cells and strongly expressed on EBV-transformed B lymphoblasts. Functionally, CD23 is involved in B cell growth and differentiation, and IgE production. Further, CD23 has a soluble form that is a potent mitogenic factor. Diseases associated with CD23 dysfunction include chronic conjunctivitis and chronic lymphocytic leukemia.
Specifications
Specifications
FCER2
Monoclonal
Unconjugated
FCER2
Lyophilized
FCER2
Human recombit protein fragment corresponding to amino acids 48-321 of human FCER2 produced in SF9 cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI9B6
PBS with 8% trehalose and no preservative; pH 7.3
P06734
BLAST-2, CD23, CD23A, CLEC4J, FCE2, IGEBF
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
2208