Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FcgammaRII/CD32 Mouse anti-Human, Janelia Fluor 549, Clone: MM0270- 3L1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP212161JF549
Description
Fc gamma RII/CD32 Monoclonal specifically detects Fc gamma RII/CD32 in Human samples. It is validated for Flow Cytometry.Specifications
FcgammaRII/CD32 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FCGR2B | |
Human recombinant Fc gamma RII/CD32 (extracellular domain) | |
0.1 mL | |
Primary | |
Detects human Fc gamma RIIa (CD32a) and Fc gamma RIIb (CD32b) | |
Store at 4°C in the dark. | |
IgG2 |
Western Blot, Flow Cytometry, Immunocytochemistry | |
MM0270- 3L1 | |
Western Blot, Flow Cytometry, Immunocytochemistry | |
CD32 antigen, CD32B, CD32Fc fragment of IgG, low affinity II, receptor for (CD32), CDw32, Fc fragment of IgG, low affinity IIb, receptor (CD32), Fc fragment of IgG, low affinity IIb, receptor for (CD32), Fc gamma RIIb, FCG2, Fc-gamma RII-b, fc-gamma-RIIb, FCGR2, fcRII-b, IGFR2, IgG Fc receptor II-b, low affinity immunoglobulin gamma Fc region receptor II-b | |
Mouse | |
Protein G purified | |
Immunology | |
2213 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title