Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FCGR2A (CD32a) Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

Manufacturer:  Sino Biological™ 10374H08B250

 View more versions of this product

Catalog No. 10-374-H08B25


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

  • Lyophilized in 20mM Tris buffer, 50mM NaCl, pH 7.4
  • Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Description & Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
2212
≥85% by SDS-PAGE
Human
<1 EU/μg
FCGR2A
CD Proteins
FCGR2A (CD32a) Recombinant Human Protein
Lyophilized
22kd
5 x 50μg
Truncated, Recombinant, Cell Surface Protein
Fc gamma receptor II-a
Assay Standard
Insect
His Tag
Provide Content Correction

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit