Sino Biological™ FCGR2A (CD32a) Recombinant Human Protein Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

Manufacturer: Sino Biological™ 10374H08B50

 View more versions of this product

Catalog No. 10-374-H08B50

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name Fc gamma receptor II-a
    Structural Form Truncated, Recombinant, Cell Surface Protein
    Molecular Weight 22kd
    Purity ≥85% by SDS-PAGE
    Source Human
    Validated Application Assay Standard
    Expression System Insect
    Product Type FCGR2A (CD32a) Recombinant Human Protein
    Gene ID (Entrez) 2212
    Quantity 50μg
    Endotoxin Concentration <1 EU/μg
    Gene Symbol FCGR2A
    Protein Tag His Tag
    Protein Family CD Proteins
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized

    Expressed from DNA sequence encoding extracellular domain (Met 1-Gly 216) of human FCGR2A precursor fused to polyhistidine tag at C-terminus

    • Lyophilized in 20mM Tris buffer, 50mM NaCl, pH 7.4
    • Contains 5% Mannitol, 5% Trehalose, and 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit