Sino Biological™ FCGR2B (CD32b) Recombinant Human Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Ala 46-Pro 217) of human FCGR2B w/N-terminal signal peptide, C-terminal polyhistidine tag

Manufacturer: Sino Biological™ 10259H08H5

 View more versions of this product

Catalog No. 10-259-H08H5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Molecular Weight 20.8kd
    Purity ≥90% by SDS-PAGE
    Quantity 5μg
    Source Human
    Storage Requirements −20° to −70°C
    Endotoxin Concentration <1 EU/μg
    Common Name Fc gamma receptor II-b
    Gene Symbol FCGR2B
    Protein Tag His Tag
    Validated Application Assay Standard
    Protein Family CD Proteins, Growth Factors and Receptors
    Expression System Mammalian
    For Use With (Application) Analysis of protein structure, studying protein-protein interactions and cell biology research
    Format Lyophilized
    Gene ID (Entrez) 2213
    Structural Form Truncated, Recombinant, Cell Surface Protein
    Product Type FCGR2B (CD32b) Recombinant Human Protein

    Expressed from DNA sequence encoding extracellular domain (Ala 46-Pro 217) of human FCGR2B w/N-terminal signal peptide, C-terminal polyhistidine tag

    • Measured by its binding ability in functional ELISA
    • Immobilized recombinant human FCGR2B at 10µg/mL binds human IgG2 w/linear range of 0.16-6.4µg/mL
    • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit