Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fcgr3, anti-Mouse, Clone: 2.4G2, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00112

Catalog No. 89-032-455


Add to cart

Specifications

Specifications

Fcgr3
2.4G2
Blocking Flow Cytometry (1 ug/mL) Immunofluorescence Immunohistochemistry (5 ug/mL) Immunoprecipitation (10 ug/mL) The optimal working dilution should be determined by the end user.
Fc receptor, IgG, low affinity III
CD16
Original antibody is raised against mouse BALB/c macrophage J774 cell line.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
14131|14130
Mouse
Blocking Assay, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Liquid
Fcgr3
IgG, kappa
RUO
Antibody
Monoclonal
This antibody binds to an epitope on the extracellular domains of the mouse Fc gamma III and Fc gamma II receptors, thereby inhibiting their ability to bind antibodies via their Fc region. Also binding the Fc gamma I receptor (CD64) via its Fc domain.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit