Learn More
FCGR3A 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for FCGR3A
Manufacturer: Abnova Corporation H00002214T01
Description
This gene encodes a receptor for the Fc portion of immunoglobulin G, and it is involved in the removal of antigen-antibody complexes from the circulation, as well as other other antibody-dependent responses. This gene (FCGR3A) is highly similar to another nearby gene (FCGR3B) located on chromosome 1. The receptor encoded by this gene is expressed on natural killer (NK) cells as an integral membrane glycoprotein anchored through a transmembrane peptide, whereas FCGR3B is expressed on polymorphonuclear neutrophils (PMN) where the receptor is anchored through a phosphatidylinositol (PI) linkage.
- Fc fragment of IgG, low affinity IIIa, receptor (CD16a)
- Transfected Cell Line: 293T
- Plasmid: pCMV-FCGR3A full-length
- Host: Human
- Theoretical MW: 32.6kDa
- Storage Buffer: 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Western Blotting
Specifications
Western Blot | |
FCGR3A 293T Cell Transient Overexpression Lysate(Denatured) | |
32.6 | |
pCMV-FCGR3A full-length | |
100 μL | |
Human |
293T | |
BC036723.1 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-FCGR3A antibody (H00002214-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.