Sino Biological™ FCGR3A (CD16a), 158 Phe/Val Recombinant Human Protein, His.AVI Tag, Biotinylated Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A w/176 Phe/Val mutation

Manufacturer: Sino Biological™ 10389H27HB5

 View more versions of this product

Catalog No. 10-389-H27HB5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name Fc gamma receptor III-a
    Structural Form Truncated, Recombinant, Cell Surface Protein
    Protein Tag AVI Tag, His Tag
    Protein Family CD Proteins, Growth Factors and Receptors
    Molecular Weight 25.6kd
    Purity ≥85% by SDS-PAGE
    Quantity 5μg
    Source Human
    Endotoxin Concentration <1 EU/μg
    Gene Symbol FCGR3A
    Validated Application Assay Standard
    Expression System Mammalian
    Product Type FCGR3A (CD16a), 158 Phe/Val Recombinant Human Protein
    For Use With (Application) Studying protein-protein interactions and cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 2214

    Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A w/176 Phe/Val mutation

    • Used to polyhistidine and AVI tags at C-terminus
    • Expressed protein was biotinylated in vivo by Biotin-Protein ligase (BirA enzyme) which co-expressed
    • Measured by its ability to bind recombinant human IgG1 in functional ELISA
  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit