Sino Biological™ FCGR3A (CD16a) Recombinant Human Protein, His.AVI Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A containing 176 Phe/Val mutation fused to a polyhistidine tag

Manufacturer: Sino Biological™ 10389H27H250

 View more versions of this product

Catalog No. 10-389-H27H25

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Common Name Fc gamma receptor III-a
    Structural Form Truncated, Recombinant, Cell Surface Protein
    Protein Family CD Proteins, Growth Factors and Receptors
    Molecular Weight 23.3kd
    Purity ≥90% by SDS-PAGE
    Endotoxin Concentration <1 EU/μg
    Protein Tag His.AVI Tag
    Validated Application Assay Standard
    Expression System Mammalian
    Product Type FCGR3A (CD16a) Recombinant Human Protein
    For Use With (Application) Studying protein-protein interactions, and cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 2214
    Quantity 5 x 5μg
    Source Human
    Gene Symbol FCGR3A (CD16a)

    Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 208) of human FCGR3A containing 176 Phe/Val mutation fused to a polyhistidine tag

    • Measured using Octet RED System affinity constant (Kd) of FCGR3A bound to human IgG1 was 80nM
    • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit