Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCGR3A Mouse anti-Human, Clone: 3G8 (Biotin), Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB13820

Catalog No. 89-033-885


Add to cart

Description

Description

Mouse monoclonal antibody raised against human FCGR3A.

  • High specificity
  • High reactivity
Specifications

Specifications

FCGR3A
3G8
Mouse monoclonal antibody raised against human FCGR3A.
In PBS, pH 7.4 (protein stabilizer, 0.09% sodium azide).
Liquid
FCGR3A
Human polymorphonuclear leukocytes.
Protein A/G purification
RUO
Primary
2214
Flow Cytometry, Functional Assay, Immunohistochemistry (Frozen), Immunoprecipitation
Biotin
Flow Cytometry (1 ug/106 cells) Functional Study Immunohistochemistry (Frozen sections) Immunoprecipitation The optimal working dilution should be determined by the end user.
P08637
CD16|CD16A|FCG3|FCGR3|FCGRIII|FCR-10|FCRIII|FCRIIIA|IGFR3
Mouse
IgG1
100 ug
Store in the dark at 4°C. Avoid prolonged exposure to light.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)