Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sino Biological™ FCGR4 (CD16-2) Recombinant Mouse Protein, His Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 203) of mouse FCGR4 fused to polyhistidine tag at C-terminus

Manufacturer:  Sino Biological™ 50036M08H5

 View more versions of this product

Catalog No. 50-036-M08H5


  Check Availability
Add to cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Expressed from DNA sequence encoding extracellular domain (Met 1-Gln 203) of mouse FCGR4 fused to polyhistidine tag at C-terminus

  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
Description & Specifications

Specifications

Analysis of protein stucture and used as immunogen, protein standard, or in cell biology research applications
246256
≥90% by SDS-PAGE
Mouse
<1 EU/μg
Fcgr4
CD Proteins
FCGR4 (CD16-2) Recombinant Mouse Protein
Lyophilized
22.4kd
5μg
Truncated, Cell Surface Protein, Recombinant
Fc gamma receptor receptor IV
Assay Standard
Mammalian
His Tag
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit