Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FCP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530302
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (97%), Rat (95%), Zebrafish (95%), Xenopus laevis (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein which interacts with the carboxy-terminus of transcription initiation factor TFIIF, a transcription factor which regulates elongation as well as initiation by RNA polymerase II. The protein may also represent a component of an RNA polymerase II holoenzyme complex. Alternative splicing of this gene results in two transcript variants encoding 2 different isoforms.
Specifications
FCP1 | |
Polyclonal | |
Unconjugated | |
CTDP1 | |
4930563P03Rik; AW553592; CCFDN; CTD (carboxy-terminal domain, RNA polymerase II, polypeptide A) phosphatase, subunit 1; CTD (carboxy-terminal domina, RNA polymerase II, polypeptide A) phosphatase, subunit 1; CTD of POLR2A, phosphatase of, subunit 1; CTD phosphatase subunit 1; Ctdp1; FCP1; RNA polymerase II subunit A C-terminal domain phosphatase; serine phosphatase FCP1a; TFIIF-associating CTD phosphatase; TFIIF-associating CTD phosphatase 1; transcription factor IIF-associating CTD phosphatase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9150 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9Y5B0 | |
CTDP1 | |
Recombinant fragment corresponding to a region within amino acids 81 and 309 of Human FCP1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction