Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FCRL4/FcRH4/IRTA1 Mouse anti-Human, Alexa Fluor 594, Clone: A1, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311836AF594
Description
FCRL4/FcRH4/IRTA1 Monoclonal antibody specifically detects FCRL4/FcRH4/IRTA1 in Human samples. It is validated for Flow CytometrySpecifications
FCRL4/FcRH4/IRTA1 | |
Monoclonal | |
Alexa Fluor 594 | |
CD307d antigen, Fc receptor homolog 4, Fc receptor-like 4, FcRH4, FCRH4Fc receptor-like protein 4, FcRL4, FcR-like protein 4, hIFGP2, IFGP family protein 2, IFGP2, IGFP2, Immune receptor translocation-associated protein 1, immunoglobulin superfamily Fc receptor, gp42, immunoglobulin superfamily receptor translocation associated 1, IRTA1MGC150523, MGC150522 | |
DNA-immunization followed by a boost with FCRL4/FcRH4/IRTA1-transfected cells | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
A1 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
83417 | |
Store at 4°C in the dark. | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title