Learn More
FCRLA/FCRLM1 Rat anti-Mouse, Alexa Fluor 750, Clone: 865910, R&D Systems™
Rat Monoclonal Antibody
Manufacturer:  R&D Systems FAB7905S100UG
Catalog No. FAB7905S100
Add to cart
This item is not returnable. View return policy
Description
Description
FCRLA/FCRLM1 Monoclonal specifically detects FCRLA/FCRLM1 in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
Specifications
FCRLA/FCRLM1
Monoclonal
0.2 mg/mL
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Q920A9
Fc receptor homolog expressed in B cells (FREB), Fc receptor homolog expressed in B-cells, Fc receptor related protein X, Fc receptor-like A, Fc receptor-like and mucin-like 1, Fc receptor-related protein X, FCRL1, FCRLa, FCRLb, FCRLc1, FCRLc2, FCRLd, FCRLe, FCRLFc receptor-like protein, FCRLM1, FCRLX, FCRX, FREB, FREBFc receptor-like and mucin-like protein 1, MGC4595
Rat
IgG2a
100 ug
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Monoclonal
Detects mouse FCRLA in ELISA.
Flow Cytometry
865910
Alexa Fluor 750
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Purified
FCRLA
E. coli-derived recombinant mouse FCRLA Lys221-Lys352 Accession # Q920A9
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
84824
Mouse
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)