Learn More
FCRLB Mouse anti-Human, Clone: OTI5H5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809246
Catalog No. 50-166-9367
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
FCRL2 belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions.
Specifications
Specifications
FCRLB
Monoclonal
Unconjugated
FCRLB
Lyophilized
FCRLB
Human recombit protein fragment corresponding to amino acids 202-426 of human FCRLB produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI5H5
PBS with 8% trehalose and no preservative; pH 7.3
Q6BAA4
FCRL2, FCRLM2, FCRLY, FREB-2, FREB2, FcRY
Mouse
IgG2b
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
127943