Learn More
Mouse anti-FCRN/FCGRT, DyLight 650, Clone: 2E9.1G8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242214C
Catalog No. NBP242214C
Add to cart
Description
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
FCRN/FCGRT Monoclonal antibody specifically detects FCRN/FCGRT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
FCRN/FCGRT
2E9.1G8
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
P55899
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Mouse
IgG2b Kappa
Protein G purified
RUO
Primary
2217
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DyLight 650
50mM Sodium Borate with 0.05% Sodium Azide
Purified
FCGRT
Partial recombinant human FCGRT protein (between amino acids 10-200) [Uniprot: P55899]
39.7 kDa
0.1 ml
Store at 4C in the dark.
Monoclonal
Human