Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRN/FCGRT Mouse anti-Human, Clone: FCGRT/2932, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30722810UG

 View more versions of this product

Catalog No. NB121929


Add to cart

Description

Description

FCRN/FCGRT Monoclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

FCRN/FCGRT
FCGRT/2932
Unconjugated
10 mM PBS with 0.05% BSA
alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Recombinant fragment of human FCRN/FCGRT protein (around aa 24-215) (Exact sequence is proprietary)
Protein A or G purified
RUO
Primary
2217
Immunohistochemistry (Paraffin), Peptide Array
0.2 mg/ml
Immunohistochemistry-Paraffin 2-4 ug/ml, Protein Array
Purified
Mouse
IgG2b Kappa
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit