Learn More
ACROBiosystems Recombinant Protein;Biotinylated Bovine FcRn / FCGRT & B2M Heterodimer Protein;HEK293;25UG;FCN,FcRN,FCGRT & B2M,FCGRT&B2M,B2M,Beta-2 microglobulin,Fcr,Fcgr,fc gamma,fc receptor,Fc gamma receptor
Supplier:  ACROBiosystems FCNB82W325UG
Encompass_Preferred
Recombinant Protein;Biotinylated Bovine FcRn / FCGRT & B2M Heterodimer Protein;25UG;MABSol Biotinylated Bovine FcRn Heterodimer Protein (FCN-B82W3) is expressed from human HEK293 cells. It contains AA Ala 24 - Pro 298 (FCGRT) & Ile 21 - Leu 118 (B2M) (Accession # Q3T119-1 (FCGRT) & P01888-1 (B2M)).;The subunit FCGRT carries a polyhistidine tag at the C-terminus, followed by an Avi tag (Avitag™). The protein has a calculated MW of 34.5 kDa (FCGRT), 11.6 kDa (B2M) . The protein migrates as 37 kDa (FCGRT), 12 kDa (B2M) respectively on a SDS-PAGE gel under reducing (R) condition due to glycosylation.;Immobilized Ipilumimab at 2 g/mL (100 L/well) can bind Biotinylated Bovine FcRn Heterodimer Protein (Cat. No. FCN-B82W3) with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-165-7698