Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Alfa Aesar™ FCS Blocking Buffer, in PBS
For blocking excess solid surface in ELISA assays
Manufacturer:  Alfa Aesar™ J63160AK
 View more versions of this product
Catalog No. AAJ63160AK
Order in Bulk
Request bulk or custom formats
Specifications
Specifications
For blocking excess solid surface in ELISA assays
10% Fetal calf serum in PBS
Blocking Buffer
10% Fetal calf serum in PBS, 0.8% Sodium chloride, 0.144% Sodium hydrogen phosphate, 0.024% Potassium dihydrogen phosphate, 0.02% Potassium chloride, 89.012% Water
Liquid
250mL