Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCS Blocking Buffer in TBS

For blocking excess solid surface in ELISA

$54.80 - $95.90

Specifications

Format Liquid
For Use With (Application) For blocking excess solid surface in ELISA
Product Type Blocking Buffer
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ61327AK View Documents Alfa Aesar™
J61327AK
250 mL Each for $54.80
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAJ61327AP View Documents Alfa Aesar™
J61327AP
500 mL Each for $95.90
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Liquid
For blocking excess solid surface in ELISA
Blocking Buffer
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit