Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Carmine, Spectrum™

Click to view available options
Quantity:
10 g
25 g
100 g
500 g
Packaging:
Amber Glass Bottle
1390-65-4
C22H20O13
492.39
DGQLVPJVXFOQEV-UHFFFAOYNA-N
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O
1390-65-4
C22H20O13
492.39
DGQLVPJVXFOQEV-UHFFFAOYNA-N
3,5,6,8-tetrahydroxy-1-methyl-9,10-dioxo-7-[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracene-2-carboxylic acid
CC1=C(C(O)=O)C(O)=CC2=C1C(=O)C1=C(O)C(C3OC(CO)C(O)C(O)C3O)=C(O)C(O)=C1C2=O

F1012, 1390-65-4, C22H20O13

Spectrum™ FD and C Carmine
CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g