Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Green No. 3, Spectrum™ Chemical
SDP

Catalog Number 18602622
Click to view available options
Quantity:
25 g
100 g
Packaging:
Amber Glass Bottle
2353-45-9
C37H34N2Na2O10S3
808.84
MFCD00013053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
2353-45-9
C37H34N2Na2O10S3
808.84
MFCD00013053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

FD120, 2353-45-9, C37H34N2O10S3Na2

Spectrum™ Chemical FD and C Green No. 3
CAS Max % 100%
Grade Reagent
Packaging Amber Glass Bottle
Quantity 25 g
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit