Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FD and C Green No. 3, Spectrum™

Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
2353-45-9
C37H34N2Na2O10S3
808.84
XJBPDZVCYTYRIN-UHFFFAOYSA-L
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
2353-45-9
C37H34N2Na2O10S3
808.84
XJBPDZVCYTYRIN-UHFFFAOYSA-L
disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(4-hydroxy-2-sulfonatophenyl)methylium
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O

FD120, 2353-45-9, C37H34N2O10S3Na2

Spectrum™ FD and C Green No. 3
CAS Max % 1
Packaging Poly Pail
Quantity 5 kg