Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FDCSP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550662
Description
The antibody detects endogenous levels of total FDCSP protein.
FDCSP Polyclonal antibody specifically detects FDCSP in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
FDCSP | |
Polyclonal | |
Unconjugated | |
FDCSP | |
C4orf7; FDC secreted protein; FDCSP; FDC-SP; follicular dendritic cell secreted; follicular dendritic cell secreted peptide; follicular dendritic cell secreted protein; LOC494224; UNQ733/PRO1419 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
260436 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8NFU4 | |
FDCSP | |
Synthetic peptide corresponding to a region derived from internal residues of human follicular dendritic cell secreted protein | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction