Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FE65 Mouse anti-Mouse, Human, Clone: 3H6, MilliporeSigma™

Mouse Monoclonal Antibody

$155.70 - $367.50

Specifications

Antigen FE65
Clone 3H6
Applications Western Blot
Host Species Mouse
Target Species Mouse, Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
05-758-I25UG
View Documents
MilliporeSigma
05758I25UG
25 μg
Each for $155.70
Add to cart
 
05-758-I100UG
View Documents
MilliporeSigma
05758I100UG
100 μg
Each for $367.50
Add to cart
 
Description

Description

Anti-FE65, clone 3H6 is a highly specific mouse monoclonal antibody that targets FE65 and has been tested for use in Western Blotting.
Specifications

Specifications

FE65
Western Blot
Mouse, Human
Primary
3H6
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit