Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FE65 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP303692100UL

 View more versions of this product

Catalog No. NB093105


Add to cart

Description

Description

FE65 Polyclonal antibody specifically detects FE65 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

FE65
Unconjugated
Purified
Rabbit
IgG
100 μL
Alzheimers Research, Cell Cycle and Replication, Neurodegeneration, Neuroscience, Phospho Specific, Signal Transduction
Primary
322
Immunofluorescence, Immunohistochemistry, Western Blot
PBS with 50% glycerol, pH7.3.
adaptor protein FE65a2, amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65), amyloid beta A4 precursor protein-binding, family B, member 1, Fe65, FE65amyloid beta A4 precursor protein-binding family B member 1, MGC:9072, Protein Fe65, RIR, stat-like protein
Recombinant fusion protein containing a sequence corresponding to amino acids 359-708 of human FE65 (NP_663722.1).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit