Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LW Scientific USA E8 Centrifuge Products for Science Education

Features auto-brake to stop rotor in less than 30 seconds.

Manufacturer:  LW Scientific E8CU8AV1503/EMD

Catalog No. S14187


Add to cart

Description

Description

  • Accurate speeds for specific g-forces
  • Quiet, vibration-free operation
  • Angled rotor, 8-tube capacity
  • Holds 3mL to 15mL size tubes
  • Maintenance-free motor 12V DC
  • Proper separations without cell damage
  • Clear view port in lid for using tachometer
  • Suction-cupped feet prevent slipping
  • Auto-switching power adapter
  • Lid safety shut-off switch
Specifications

Specifications

24.1 x 27.9 x 27.9 cm (9.5 x 11 x 11 in.)
AC-adapter power cord, large 15mL tube sleeves, small 4mL tube sleeve inserts
110/220 V
Variable Speed
9.5 lb. (4.3 kg)
3,500 rpm
ISO:9001-13485, CE, CSA (CE, UL, cUL approved)
Spins blood, urine, fecals, semen
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit