Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Federal Signal Pulsator Strobe Light Non-distribution product as customer accommodation.

Manufacturer:  Federal Signal 21168102

 View more versions of this product

Catalog No. 19-820-002


Add to cart

Specifications

Specifications

1.2 to 0.3 A
Amber
6.5 in.
Strobe Beacon
15.2cm
6 in.
13.3w
12/48VDC
16.5cm
SAE J845 Class 2, J1318 Class 2
Forklifts, industrial trucks, construction vehicles
Polycarbonate
Permanent
80 fpm
Safety and Handling

Safety and Handling

ShelfLife : 3 Years

Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit