Learn More
Federal Signal VisiBeam Spotlight
Supplier:  Federal Signal 620142
Encompass
Catalog No. 19-820-036
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.
Description
Description
Specifications
Specifications
Spotlight
100 W
6.5 in.
6.9 in.
Magnetic mount and cigarette lighter plug with 10 ft. cord
12 VDC
7.8 A
16.5 cm
17.5 cm
Magnetic mount with cigarette plug