Learn More
Fehling's Alkaline Solution (Fehling Solution B), Ricca Chemical
Manufacturer:  Ricca Chemical CompanySupplier Diversity Partner 30001
Packaging Poly Bottle
Quantity 4L
 View more versions of this product
Catalog No. 3000-1
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
1310-73-2 (Sodium Hydroxide),6381-59-5 (Sodium Potassium Tartrate Tetrahydrate),7732-18-5 (Water, Deionized)
Liquid
Fehling's Alkaline Solution, Fehling Solution B
Specifications
1310-73-2 (Sodium Hydroxide),6381-59-5 (Sodium Potassium Tartrate Tetrahydrate),7732-18-5 (Water, Deionized)
Poly Bottle
4L
Liquid
Clear colorless
Fehling's Alkaline Solution, Fehling Solution B