Learn More
Fehling's Copper Solution (Fehling Solution A), Ricca Chemical
For Determination of Reducing Sugars
Supplier: Ricca Chemical Company 30105
Specifications
Fehling's Copper Solution | |
Fehling Solution A | |
Cubitainer | |
Liquid |
Blue | |
7758-99-8, 7732-18-5 | |
20 L |