Learn More
Sklar InstrumentsSupplier Diversity Partner Feilchenfeld Splinter Forceps, Sklar, 4-1/2 in., Sharp, Straight, Serrated with Groove, Stainless Steel. Sklar Feilchenfeld Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 192945
Feilchenfeld Splinter Forceps, Sklar, 4-1/2 in., Sharp, Straight, Serrated with Groove, Stainless Steel. Sklar Feilchenfeld Splinter Forceps are multi-purpose thumb forceps with sharp tips used for removing small foreign objects. They feature a wide thumb grip on the handle for increased control and precision. This product is straight with serrated tips, a central groove, and a length of 4-1/2 inches.
Catalog No. 50-143-7433