Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Felbamate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
25451-15-4
C11H14N2O4
238.243
WKGXYQFOCVYPAC-UHFFFAOYSA-N
felbamate,felbatol,2-phenylpropane-1,3-diyl dicarbamate,2-phenyl-1,3-propanediol dicarbamate,felbamyl,taloxa,felbamato,felbamatum,felbamatum latin,felbamato spanish
3331
CHEBI:4995
(3-carbamoyloxy-2-phenylpropyl) carbamate
C1=CC=C(C=C1)C(COC(=O)N)COC(=O)N
25451-15-4
C11H14N2O4
238.243
WKGXYQFOCVYPAC-UHFFFAOYSA-N
felbamate,felbatol,2-phenylpropane-1,3-diyl dicarbamate,2-phenyl-1,3-propanediol dicarbamate,felbamyl,taloxa,felbamato,felbamatum,felbamatum latin,felbamato spanish
3331
CHEBI:4995
(3-carbamoyloxy-2-phenylpropyl) carbamate
C1=CC=C(C=C1)C(COC(=O)N)COC(=O)N

NMDA antagonist, acts glycine site

Anticonvulsant, acting as an antagonist at the NMDA-associated glycine binding site.
Quantity 50mg
Formula Weight 238.24
Percent Purity >99%
Chemical Name or Material Felbamate

Recommended Storage : Store at Room Temperature