Learn More
Raybiotech IncSupplier Diversity Partner Feline Galectin-3 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELFGALECTIN31
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 4.5 pg/mL. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Feline Galectin-3.
Catalog No. 50-194-8915