Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Herpes Virus Type 1 Antibody (FHV7-7C), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063151AF750
Description
Feline Herpes Virus Type 1 Monoclonal antibody specifically detects Feline Herpes Virus Type 1 in Virus samples. It is validated for Western Blot, ELISASpecifications
Feline Herpes Virus Type 1 | |
Monoclonal | |
Alexa Fluor 750 | |
Mouse | |
Protein A purified | |
RUO | |
Virus | |
Purified |
Western Blot, ELISA | |
FHV7-7C | |
50mM Sodium Borate | |
Made to Viral FELINE HERPES VIRUS TYPE 1 | |
0.125 mL | |
Primary | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Product Content Correction