Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Feline Herpes Virus Type 1 Antibody (FHV7-7C) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB100631510.025MG
Description
Feline Herpes Virus Type 1 Monoclonal specifically detects Feline Herpes Virus Type 1 in Virus samples. It is validated for Western Blot, ELISA.Specifications
Feline Herpes Virus Type 1 | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Mouse | |
Protein A purified | |
RUO | |
NB100-63151 is specific for feline Herpesvirus type 1 (FHV-1), of the genus Herpesviridae. FHV-1 is worldwide, and the most clinically significant pathogen for feline respiratory infections, being the causative agent of feline viral rhinotracheitis (FVR). Clone FHV7-7C specifically reacts with three different cultured FHV isolates, but does not cross react with feline Calcivirus (FCV), feline Immunodeficiency virus (FIV) or feline Leukaemia virus (FeLV). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
ELISA, Western Blot | |
FHV7-7C | |
Unconjugated | |
Phosphate buffered saline with 0.05% Sodium Azide | |
Made to Viral FELINE HERPES VIRUS TYPE 1 | |
0.025 mg | |
Primary | |
Virus | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction