Learn More
Feline IgG, F(ab)2, Biotin, Isotype control, Novus Biologicals™
Feline Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197095
Catalog No. NBP197095
Add to cart
Description
Description
IgG F(ab)2 antibody specifically detects IgG F(ab)2 in Feline samples. It is validated for Immunocytochemistry/Immunofluorescence, Fluorophore-linked immunosorbent assay.
Specifications
Specifications
IgG F(ab)2
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Feline
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG2
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Feline IgG, anti-Feline IgG F(ab)2 and anti-Feline Serum. No reaction was observed against anti-Feline IgG F(c) or anti-Pepsin.
Feline