Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Feline IgG, F(ab), Biotin, Isotype control, Novus Biologicals™

Feline Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197096

Catalog No. NBP197096


Add to cart

Description

Description

IgG F(ab) antibody specifically detects IgG F(ab) in Feline samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

IgG F(ab)
Biotin
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Feline
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
LYOPH
Restore with 1.0mL of deionized water (or equivalent).
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Feline IgG, anti-Feline IgG F(ab)2 and anti-Feline Serum. No reaction was observed against anti-Feline IgG F(c) or anti-Papain.
Feline
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit