Learn More
Novus Biologicals™ Feline IgG, Fab2 Native Protein
Highly purified. Generating reliable and reproducible results.
Manufacturer:  Novus Biologicals™ NBP1970271MG
Catalog No. NBP1970271
Add to cart
Description
Description
Native Feline IgG, Fab2. The Feline IgG, Fab2 Native Protein has been validated for the following applications: Western Blot, ELISA.
Specifications
Specifications
1.0 mg/ml
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2
1 mg
RUO
Feline
Western Blot, ELISA
Immunoelectrophoresis (IEP)
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Feline IgG, Fab2