Learn More
Feline leukemia virus p27 Mouse anti-Virus, Clone: HM190, Abnova™
Mouse Monoclonal Antibody
Manufacturer:  Abnova Corporation MAB13549
Catalog No. 89-033-512
Add to cart
Description
Description
Mouse monoclonal antibody raised against feline leukemia virus p27.
  • High specificity
  • High reactivity
Specifications
Specifications
Feline leukemia virus p27
HM190
Mouse monoclonal antibody raised against feline leukemia virus p27.
In PBS, pH 7.2 (0.09% sodium azide).
Mouse
Protein A purification
RUO
Primary
This antibody reacts with p27 of feline leukemia virus.
ELISA, Western Blot
Unconjugated
ELISA Western Blot The optimal working dilution should be determined by the end user.
Liquid
IgG2b, kappa
500 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Virus
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)