Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Felodipine, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
5 kg
25 kg
Packaging:
Fiber Drum
Poly Pail
72509-76-3
C18H19Cl2NO4
384.25
RZTAMFZIAATZDJ-UHFFFAOYNA-N
3-ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(Cl)=C1Cl)C(=O)OC
72509-76-3
C18H19Cl2NO4
384.25
RZTAMFZIAATZDJ-UHFFFAOYNA-N
3-ethyl 5-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
CCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(Cl)=C1Cl)C(=O)OC

F1206, 72509-76-3, C18H19Cl2NO4

Spectrum™ Felodipine, USP is known as a calcium channel blocker used to treat high blood pressure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Pail
Loss on Drying 0.005
Ignition Residue 0.001
Quantity 5 kg
CAS Max % 1
Percent Purity 98 to 102%