Learn More
Immunology Consultants Feline Leukemia Virus P27 Protein (FeLV p27), 0.1 mg
SDP
Supplier:  Immunology Consultants AG200281Z
Encompass_Preferred
Recombinant FeLV p27 (aa 272-519) with 6xHIS tag was purified from E.coli. The protein has a molecular weight of 33 KDa. The product is determined to be >90% pure based on SDS-PAGE analysis. It reacts with the monoclonal Mouse anti-Feline Leukemia virus, clone CM1 (MFLV-20M-CM1) as verified by Western Blot. Optimal working dilutions should be determined experimentally by the investigator.
Catalog No. 50-224-9029
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.