Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FEM1A Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566607
Description
Immunogen sequence: VEYLIQEQPGQ EQVAGGEAQP GLPQEDPSTS QGCAQPQGAP CCSSSPEEPL NGESYESCCP TSREAAVEA Highest antigen sequence identity to the following orthologs - mouse 57%, rat 56%.
FEM1A (FEM-1 homolog a), also known as EPRAP (Prostaglandin E receptor 4-associated protein), is a 669 amino acid cytoplasmic protein that belongs to the FEM-1 family. FEM1A is present in atheromata and in macrophages derived from peripheral blood monocytes. FEM1A may be a component of an E3 ubiquitin-protein ligase complex and functions as a substrate recognition subunit. It is suggested that FEM1A participates in anti-inflammatory signaling of macrophages via its interaction with EP4. FEM1A is down-regulated in Rhabdomyosarcoma, the most common soft tissue neoplasm of children. FEM1A is considered a candidate gene for polycystic ovary syndrome.
Specifications
FEM1A | |
Polyclonal | |
Unconjugated | |
FEM1A | |
AW611390; DKFZp762M136; EPRAP; fem-1 homolog A; fem-1 homolog a (C. elegans); Fem1a; Fem1aa; FEM1a-A; FEM1-alpha; FEM1-alpha-A; feminization 1 homolog a; feminization 1 homolog a (C. elegans); prostaglandin E receptor 4-associated protein; Protein fem-1 homolog A; protein fem-1 homolog A-A; similar to fem-1 homolog a (C.elegans) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55527 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BSK4 | |
FEM1A | |
Recombinant Human FEM1A. Recombinant protein control fragment (Product #RP-107265). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction