Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FEM1C Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA517940
Description
This antibody is predicted to react with bovine, canine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Probable component of an E3 ubiquitin-protein ligase complex, in which it may act as a substrate recognition subunit.
Specifications
FEM1C | |
Polyclonal | |
Unconjugated | |
FEM1C | |
2610312A07Rik; 3632443A22Rik; AI646567; EUROIMAGE686608; EUROIMAGE783647; fem-1 homolog C; fem-1 homolog c (C. elegans); fem-1 homolog c (C.elegans); FEM1A; FEM1c; FEM1-gamma; feminization 1 homolog a; feminization 1 homolog c; KIAA1785; Protein fem-1 homolog C; protein fem-1 homolog C pseudogene; protein fem-1 homolog C-like | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
240263, 302288, 56929 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8CEF1, Q96JP0 | |
FEM1C | |
Synthetic peptide sequence (HIPEKLETFVSLHR) corresponding to the C-terminus amino acids of FEM1C (aa 604-617). | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction